1-(6-Cyclopropylpyridin-2-yl)ethanone

Lieferant: Apollo Scientific

OR315875-1G OR315875-250MG OR315875-5G
APOSOR315875-1GEA 522 EUR
APOSOR315875-1G APOSOR315875-250MG APOSOR315875-5G
1-(6-Cyclopropylpyridin-2-yl)ethanone
1-(6-Cyclopropylpyridin-2-yl)ethanone
Formel: C₁₀H₁₁NO
Molecular Weight: 161.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18255568
CAS-Nummer: 1256817-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR