Methyl 5-bromo-4-(cyclopropyl)thiophene-2-carboxylate 95%

Lieferant: Apollo Scientific

OR315882-1G OR315882-250MG
APOSOR315882-1GEA 0 EUR
APOSOR315882-1G APOSOR315882-250MG
Methyl 5-bromo-4-(cyclopropyl)thiophene-2-carboxylate 95%
Methyl 5-bromo-4-(cyclopropyl)thiophene-2-carboxylate
Formel: C₉H₉BrO₂S
Lagertemperatur: Kühlschrank
MDL: MFCD32689352
CAS-Nummer: 2514953-04-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR