trans-2,6-Dimethylpiperidin-4-one, N-BOC protected
Lieferant: Apollo Scientific
Achtung
Synonyme: N-t-BOC-trans-2,6-Dimethyl-4-oxopiperidine, (2R,6R)-rel-tert-Butyl 2,6-dimethyl-4-oxopiperidine-1-carboxylate
OR316064-1G OR316064-250MG
APOSOR316064-1GEA 835 EUR
APOSOR316064-1G APOSOR316064-250MG
trans-2,6-Dimethylpiperidin-4-one, N-BOC protected
trans-2,6-Dimethylpiperidin-4-one, N-BOC protected
Formel: C₁₂H₂₁NO₃
Molecular Weight: 227.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19230166
CAS-Nummer: 184368-70-5
Bestellen
TECHNISCHE DATEN
Environmentally Preferable