4-Amino-5-bromopyrrolo[2,3-d]pyrimidine
Lieferant: Apollo Scientific
Synonyme:
5-Bromo-7H-pyrrolo[2,3-d]pyrimidin-4-amine
OR317025-1G
OR317025-5G
APOSOR317025-1GEA
59.6
EUR
APOSOR317025-1G
APOSOR317025-5G
4-Amino-5-bromopyrrolo[2,3-d]pyrimidine
4-Amino-5-bromopyrrolo[2,3-d]pyrimidine
Formel:
C₆H₅BrN₄ Molecular Weight: 213,035 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09835119 CAS-Nummer: 22276-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...