2-Amino-2-cyclobutylacetic acid

Lieferant: Apollo Scientific

Synonyme: α-Cyclobutylglycine

OR317072-1G OR317072-5G
APOSOR317072-1GEA 163 EUR
APOSOR317072-1G APOSOR317072-5G
2-Amino-2-cyclobutylacetic acid
2-Amino-2-cyclobutylacetic acid
Formel: C₆H₁₁NO₂
Molecular Weight: 129,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264350
CAS-Nummer: 28024-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR