7-Boc-2-hydroxy-7-azaspiro[3.5]nonane 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2-hydroxy-7-azaspiro[3.5]nonane-7-carboxylate

OR317098-5G
APOSOR317098-5GEA 82.8 EUR
APOSOR317098-5G
7-Boc-2-hydroxy-7-azaspiro[3.5]nonane 97%
7-Boc-2-hydroxy-7-azaspiro[3.5]nonane
Formel: C₁₃H₂₃NO₃
Molecular Weight: 241,327 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14584466
CAS-Nummer: 240401-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR