3-(Cbz-amino)azetidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: Benzyl azetidin-3-ylcarbamate hydrochloride

OR317159-1G OR317159-5G
APOSOR317159-1GEA 85.7 EUR
APOSOR317159-1G APOSOR317159-5G
3-(Cbz-amino)azetidine hydrochloride
3-(Cbz-amino)azetidine hydrochloride
Formel: C₁₁H₁₄N₂O₂·ClH
Molecular Weight: 242,702 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11101353
CAS-Nummer: 1203099-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR