1-Boc-3-hydroxy-3-(nitromethyl)azetidine
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 3-hydroxy-3-(nitromethyl)azetidine-1-carboxylate
OR317174-1G
OR317174-5G
APOSOR317174-1GEA
72.5
EUR
APOSOR317174-1G
APOSOR317174-5G
1-Boc-3-hydroxy-3-(nitromethyl)azetidine
1-Boc-3-hydroxy-3-(nitromethyl)azetidine
Formel:
C₉H₁₆N₂O₅ Molecular Weight: 232,234 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17015985 CAS-Nummer: 1008526-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...