1-Acetyl-3-aminoazetidine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Aminoazetidin-1-yl)ethan-1-one hydrochloride, 1-(3-Aminoazetidin-1-yl)ethanone hydrochloride

OR317198-1G OR317198-250MG
APOSOR317198-1GEA 169 EUR
APOSOR317198-1G APOSOR317198-250MG
1-Acetyl-3-aminoazetidine hydrochloride 95%
1-Acetyl-3-aminoazetidine hydrochloride
Formel: C₅H₁₀N₂O·ClH
Molecular Weight: 150.607 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23703584
CAS-Nummer: 1462921-50-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR