1-(3-Oxetanyl)-1H-pyrazole-4-boronic acid pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Oxetan-3-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

OR317258-1G OR317258-250MG
APOSOR317258-1GEA 301 EUR
APOSOR317258-1G APOSOR317258-250MG
1-(3-Oxetanyl)-1H-pyrazole-4-boronic acid pinacol ester
1-(3-Oxetanyl)-1H-pyrazole-4-boronic acid pinacol ester
Formel: C₁₂H₁₉BN₂O₃
Molecular Weight: 250.102 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD21337762
CAS-Nummer: 1339890-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR