Dimethyl bicyclo[1.1.1]pentane-1,3-dicarboxylate

Lieferant: Apollo Scientific
Achtung

OR317264-1G OR317264-5G
APOSOR317264-1GEA 139 EUR
APOSOR317264-1G APOSOR317264-5G
Dimethyl bicyclo[1.1.1]pentane-1,3-dicarboxylate
Dimethyl bicyclo[1.1.1]pentane-1,3-dicarboxylate
Formel: C₉H₁₂O₄
Molecular Weight: 184.189 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD23106317
CAS-Nummer: 115913-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR