tert-Butyl 3-(ethylamino)-8-azabicyclo[3.2.1]octane-8-carboxylate 95+%

Lieferant: Apollo Scientific

OR318033-1G OR318033-5G
APOSOR318033-1GEA 441 EUR
APOSOR318033-1G APOSOR318033-5G
tert-Butyl 3-(ethylamino)-8-azabicyclo[3.2.1]octane-8-carboxylate 95+%
tert-Butyl 3-(ethylamino)-8-azabicyclo[3.2.1]octane-8-carboxylate
Formel: C₁₄H₂₆N₂O₂
Molecular Weight: 254,368 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13290989
CAS-Nummer: 1305022-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR