Methyl 7-amino-2,3-dihydrobenzo[b][1,4]dioxine-6-carboxylate 95+%

Lieferant: Apollo Scientific
Achtung

OR318105-1G OR318105-10G OR318105-5G
APOSOR318105-1GEA 503 EUR
APOSOR318105-1G APOSOR318105-10G APOSOR318105-5G
Methyl 7-amino-2,3-dihydrobenzo[b][1,4]dioxine-6-carboxylate 95+%
Methyl 7-amino-2,3-dihydrobenzo[b][1,4]dioxine-6-carboxylate
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209,199 g/mol
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03724048
CAS-Nummer: 20197-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR