2,3-Dihydro-2-oxo-1H-benzimidazole-5-boronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Dihydro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzimidazol-2-one, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazol-2(3H)-one

OR3191-1G OR3191-250MG
APOSOR3191-1GEA 480 EUR
APOSOR3191-1G APOSOR3191-250MG
2,3-Dihydro-2-oxo-1H-benzimidazole-5-boronic acid, pinacol ester
2,3-Dihydro-2-oxo-1H-benzimidazole-5-boronic acid, pinacol ester
Formel: C₁₃H₁₇BN₂O₃
Molecular Weight: 260,097 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09260491
CAS-Nummer: 710348-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR