3-(Bromoacetyl)benzoic acid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-(2-Bromoacetyl)benzoic acid, 3-Carboxyphenacyl bromide, 2-Bromo-1-(3-carboxyphenyl)ethan-1-one
OR3200-5G
OR3200-1G
OR3200-25G
APOSOR3200-5GEA
149
EUR
APOSOR3200-5G
APOSOR3200-1G
APOSOR3200-25G
3-(Bromoacetyl)benzoic acid
3-(Bromoacetyl)benzoic acid
Formel:
C₉H₇BrO₃ Molecular Weight: 243,054 g/mol Schmelzpunkt: 150…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03789105 CAS-Nummer: 62423-73-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...