N-(2,4-Dichlorophenyl)-3-oxobutanamide 95%

Lieferant: Apollo Scientific

OR32014-1G OR32014-5G
APOSOR32014-1GEA 224 EUR
APOSOR32014-1G APOSOR32014-5G
N-(2,4-Dichlorophenyl)-3-oxobutanamide 95%
N-(2,4-Dichlorophenyl)-3-oxobutanamide
Formel: C₁₀H₉Cl₂NO₂
Molecular Weight: 246,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085383
CAS-Nummer: 17223-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR