2-[(4-Methoxyphenyl)methyl]-1,2,3,4-tetrahydroisoquinoline-1,3-dione 95%
Lieferant: Apollo Scientific
OR32078-1G
OR32078-5G
APOSOR32078-1GEA
224
EUR
APOSOR32078-1G
APOSOR32078-5G
2-[(4-Methoxyphenyl)methyl]-1,2,3,4-tetrahydroisoquinoline-1,3-dione 95%
2-[(4-Methoxyphenyl)methyl]-1,2,3,4-tetrahydroisoquinoline-1,3-dione
Formel:
C₁₇H₁₅NO₃ Molecular Weight: 281,306 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00974750 CAS-Nummer: 217493-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...