(R)-2-Phenylpiperidine 97%

Lieferant: Apollo Scientific
Achtung

OR321200-250MG OR321200-100MG OR321200-1G
APOSOR321200-250MGEA 362 EUR
APOSOR321200-250MG APOSOR321200-100MG APOSOR321200-1G
(R)-2-Phenylpiperidine 97%
(R)-2-Phenylpiperidine
Formel: C₁₁H₁₅N
Molecular Weight: 161,244 g/mol
Dichte: 0,9835 g/cm³ (25 °C)
Flammpunkt: 104 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06762593
CAS-Nummer: 58613-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR