(S)-2-Phenylpiperidine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR321201-100MG
OR321201-1G
OR321201-250MG
APOSOR321201-100MGEA
75.4
EUR
APOSOR321201-100MG
APOSOR321201-1G
APOSOR321201-250MG
(S)-2-Phenylpiperidine 95%
(S)-2-Phenylpiperidine
Formel:
C₁₁H₁₅N Molecular Weight: 161,244 g/mol Dichte: 0,9835 g/cm³ (25 °C) Flammpunkt: 104 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06762615 CAS-Nummer: 70665-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...