4-(3-Chloro-5-methoxyphenyl)piperidine hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR321218-100MG
OR321218-250MG
OR321218-1G
APOSOR321218-100MGEA
473
EUR
APOSOR321218-100MG
APOSOR321218-250MG
APOSOR321218-1G
4-(3-Chloro-5-methoxyphenyl)piperidine hydrochloride 97%
4-(3-Chloro-5-methoxyphenyl)piperidine hydrochloride
Formel:
C₁₂H₁₆ClNO·ClH Molecular Weight: 262,175 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29075579 CAS-Nummer: 1783405-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...