(3-(Piperidin-4-yl)phenyl)methanol hydrochloride 90%
Lieferant: Apollo Scientific
|
Achtung
|
OR321235-100MG
OR321235-250MG
APOSOR321235-100MGEA
328
EUR
APOSOR321235-100MG
APOSOR321235-250MG
(3-(Piperidin-4-yl)phenyl)methanol hydrochloride 90%
(3-(Piperidin-4-yl)phenyl)methanol hydrochloride
Formel:
C₁₂H₁₇NO·ClH Molecular Weight: 227,73 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29075597 CAS-Nummer: 2256060-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...