3-Bromo-1H-indazole-5-carbaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR321277-1G
OR321277-250MG
APOSOR321277-1GEA
212
EUR
APOSOR321277-1G
APOSOR321277-250MG
3-Bromo-1H-indazole-5-carbaldehyde 95%
3-Bromo-1H-indazole-5-carbaldehyde
Formel:
C₈H₅BrN₂O Molecular Weight: 225,042 g/mol Schmelzpunkt: 185…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11499015 CAS-Nummer: 1086391-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...