1-Methyl-5-(piperidin-3-yl)pyridin-2(1H)-one hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR321278-100MG OR321278-250MG
APOSOR321278-100MGEA 406 EUR
APOSOR321278-100MG APOSOR321278-250MG
1-Methyl-5-(piperidin-3-yl)pyridin-2(1H)-one hydrochloride 97%
1-Methyl-5-(piperidin-3-yl)pyridin-2(1H)-one hydrochloride
Formel: C₁₁H₁₆N₂O·ClH
Molecular Weight: 228,718 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30099278
CAS-Nummer: 1931126-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR