Benzyl methyl(pyrrolidin-3-ylmethyl)carbamate 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR321306-1G
OR321306-250MG
OR321306-5G
APOSOR321306-1GEA
429
EUR
APOSOR321306-1G
APOSOR321306-250MG
APOSOR321306-5G
Benzyl methyl(pyrrolidin-3-ylmethyl)carbamate 95%
Benzyl methyl(pyrrolidin-3-ylmethyl)carbamate
Formel:
C₁₄H₂₀N₂O₂ Molecular Weight: 248,321 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08061961 CAS-Nummer: 1245649-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...