2-(Chloromethyl)imidazo[1,2-a]pyrazine 95%

Lieferant: Apollo Scientific
Achtung

OR321314-1G OR321314-250MG
APOSOR321314-1GEA 0 EUR
APOSOR321314-1G APOSOR321314-250MG
2-(Chloromethyl)imidazo[1,2-a]pyrazine 95%
2-(Chloromethyl)imidazo[1,2-a]pyrazine
Formel: C₇H₆ClN₃
Molecular Weight: 167,596 g/mol
Schmelzpunkt: 110 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD08460080
CAS-Nummer: 57937-60-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR