tert-Butyl (3-chloro-2-formylphenyl)carbamate 97%

Lieferant: Apollo Scientific
Achtung

OR321315-1G OR321315-250MG
APOSOR321315-1GEA 241 EUR
APOSOR321315-1G APOSOR321315-250MG
tert-Butyl (3-chloro-2-formylphenyl)carbamate 97%
tert-Butyl (3-chloro-2-formylphenyl)carbamate
Formel: C₁₂H₁₄ClNO₃
Molecular Weight: 255.697 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18250357
CAS-Nummer: 1260667-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR