5-Chloro-2-(ethylsulphonyl)pyridine 97%

Lieferant: Apollo Scientific
Achtung

OR321316-1G OR321316-250MG
APOSOR321316-1GEA 116 EUR
APOSOR321316-1G APOSOR321316-250MG
5-Chloro-2-(ethylsulphonyl)pyridine 97%
5-Chloro-2-(ethylsulphonyl)pyridine
Formel: C₇H₈ClNO₂S
Lagertemperatur: Kühlschrank
MDL: MFCD14706682
CAS-Nummer: 859536-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR