8-Chloro-3-iodoimidazo[1,2-a]pyrazine 97%

Lieferant: Apollo Scientific
Achtung

OR321319-1G OR321319-250MG
APOSOR321319-1GEA 188 EUR
APOSOR321319-1G APOSOR321319-250MG
8-Chloro-3-iodoimidazo[1,2-a]pyrazine 97%
8-Chloro-3-iodoimidazo[1,2-a]pyrazine
Formel: C₆H₃ClIN₃
Molecular Weight: 279,466 g/mol
Schmelzpunkt: 179…183 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11053343
CAS-Nummer: 1049677-32-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR