Ethyl 6-amino-5-methyl-1H-indole-2-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR321321-1G OR321321-250MG
APOSOR321321-1GEA 840 EUR
APOSOR321321-1G APOSOR321321-250MG
Ethyl 6-amino-5-methyl-1H-indole-2-carboxylate 97%
Ethyl 6-amino-5-methyl-1H-indole-2-carboxylate
Formel: C₁₂H₁₄N₂O₂
Molecular Weight: 218,252 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28165861
CAS-Nummer: 1175787-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR