2-(Pyrrolidin-3-yl)pyridine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Tetrahydro-1H-pyrrol-3-ylpyridine hydrochloride, 3-(Pyridin-2-yl)pyrrolidine hydrochloride

OR321362-1G OR321362-100MG
APOSOR321362-1GEA 383 EUR
APOSOR321362-1G APOSOR321362-100MG
2-(Pyrrolidin-3-yl)pyridine hydrochloride 97%
2-(Pyrrolidin-3-yl)pyridine hydrochloride
Formel: C₉H₁₂N₂·ClH
Molecular Weight: 184.666 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13176679
CAS-Nummer: 1198416-89-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR