(S)-2-Methylpiperidine hydrochloride 97%

Lieferant: Apollo Scientific

OR321379-1G
APOSOR321379-1GEA 104 EUR
APOSOR321379-1G
(S)-2-Methylpiperidine hydrochloride 97%
(S)-2-Methylpiperidine hydrochloride
Formel: C₆H₁₃N·ClH
Molecular Weight: 135.635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26125912
CAS-Nummer: 205526-61-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR