N-(2-(Piperidin-4-yl)phenyl)acetamide hydrochloride
Lieferant: Apollo Scientific
OR321397-1G
OR321397-250MG
APOSOR321397-1GEA
0
EUR
APOSOR321397-1G
APOSOR321397-250MG
N-(2-(Piperidin-4-yl)phenyl)acetamide hydrochloride
N-(2-(Piperidin-4-yl)phenyl)acetamide hydrochloride
Formel:
C₁₃H₁₈N₂O·ClH Lagertemperatur: Kühlschrank |
MDL:
MFCD29075582 CAS-Nummer: 255050-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...