4-(3-(Methylsulphonyl)phenyl)piperidine hydrochloride
Lieferant: Apollo Scientific
OR321436-1G
OR321436-250MG
APOSOR321436-1GEA
719
EUR
APOSOR321436-1G
APOSOR321436-250MG
4-(3-(Methylsulphonyl)phenyl)piperidine hydrochloride
4-(3-(Methylsulphonyl)phenyl)piperidine hydrochloride
Formel:
C₁₂H₁₇NO₂S·ClH Lagertemperatur: Kühlschrank |
MDL:
MFCD19382377 CAS-Nummer: 346688-60-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...