3-Chloropyrazine-2-carbaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR321482-1G
OR321482-250MG
APOSOR321482-1GEA
435
EUR
APOSOR321482-1G
APOSOR321482-250MG
3-Chloropyrazine-2-carbaldehyde 97%
3-Chloropyrazine-2-carbaldehyde
Formel:
C₅H₃ClN₂O Molecular Weight: 142,543 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371660 CAS-Nummer: 121246-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...