3-Methyl-2-(piperidin-4-yl)pyridine dihydrochloride

Lieferant: Apollo Scientific

OR321490-1G OR321490-250MG
APOSOR321490-1GEA 957 EUR
APOSOR321490-1G APOSOR321490-250MG
3-Methyl-2-(piperidin-4-yl)pyridine dihydrochloride
3-Methyl-2-(piperidin-4-yl)pyridine dihydrochloride
Formel: C₁₁H₁₆N₂·2ClH
Molecular Weight: 249.18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29075595
CAS-Nummer: 1279029-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR