4-(3-Ethylphenyl)piperidine hydrochloride
Lieferant: Apollo Scientific
OR321491-1G
OR321491-250MG
APOSOR321491-1GEA
493
EUR
APOSOR321491-1G
APOSOR321491-250MG
4-(3-Ethylphenyl)piperidine hydrochloride
4-(3-Ethylphenyl)piperidine hydrochloride
Formel:
C₁₃H₁₉N·ClH Molecular Weight: 225,758 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD30099246 CAS-Nummer: 2256060-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...