(1R,3R)-3-((tert-Butoxycarbonyl)amino)cyclohexane-1-carboxylic acid

Lieferant: Apollo Scientific

OR321499-1G OR321499-250MG
APOSOR321499-1GEA 672 EUR
APOSOR321499-1G APOSOR321499-250MG
(1R,3R)-3-((tert-Butoxycarbonyl)amino)cyclohexane-1-carboxylic acid
(1R,3R)-3-((tert-Butoxycarbonyl)amino)cyclohexane-1-carboxylic acid
Formel: C₁₂H₂₁NO₄
Molecular Weight: 243.299 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD24395695
CAS-Nummer: 1430938-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR