4-(Benzo[d][1,3]dioxol-5-yl)piperidine hydrochloride
Lieferant: Apollo Scientific
OR321501-1G
OR321501-250MG
APOSOR321501-1GEA
957
EUR
APOSOR321501-1G
APOSOR321501-250MG
4-(Benzo[d][1,3]dioxol-5-yl)piperidine hydrochloride
4-(Benzo[d][1,3]dioxol-5-yl)piperidine hydrochloride
Formel:
C₁₂H₁₅NO₂·ClH Molecular Weight: 241,714 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
2409597-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...