N-(3-(Piperidin-3-yl)phenyl)acetamide hydrochloride

Lieferant: Apollo Scientific

OR321503-1G OR321503-250MG
APOSOR321503-1GEA 877 EUR
APOSOR321503-1G APOSOR321503-250MG
N-(3-(Piperidin-3-yl)phenyl)acetamide hydrochloride
N-(3-(Piperidin-3-yl)phenyl)acetamide hydrochloride
Formel: C₁₃H₁₈N₂O·ClH
Lagertemperatur: Kühlschrank
CAS-Nummer: 2409589-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR