2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-1,2,3,6-tetrahydropyridin-4-yl)benzoic acid

Lieferant: Apollo Scientific

OR321520-1G OR321520-250MG
APOSOR321520-1GEA 877 EUR
APOSOR321520-1G APOSOR321520-250MG
2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-1,2,3,6-tetrahydropyridin-4-yl)benzoic acid
2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-1,2,3,6-tetrahydropyridin-4-yl)benzoic acid
Formel: C₂₇H₂₃NO₄
Molecular Weight: 425,476 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 2416262-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR