Methyl 2-(piperidin-2-yl)benzoate hydrochloride

Lieferant: Apollo Scientific

OR321531-1G OR321531-250MG
APOSOR321531-1GEA 799 EUR
APOSOR321531-1G APOSOR321531-250MG
Methyl 2-(piperidin-2-yl)benzoate hydrochloride
Methyl 2-(piperidin-2-yl)benzoate hydrochloride
Formel: C₁₃H₁₇NO₂·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD08460734
CAS-Nummer: 1203685-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR