tert-Butyl (S)-3-(cyanomethyl)piperidine-1-carboxylate
Lieferant: Apollo Scientific
OR321533-1G
OR321533-250MG
APOSOR321533-1GEA
687
EUR
APOSOR321533-1G
APOSOR321533-250MG
tert-Butyl (S)-3-(cyanomethyl)piperidine-1-carboxylate
tert-Butyl (S)-3-(cyanomethyl)piperidine-1-carboxylate
Formel:
C₁₂H₂₀N₂O₂ Molecular Weight: 224,299 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08272979 CAS-Nummer: 1217710-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...