(S)-4-(Piperidin-3-yl)aniline

Lieferant: Apollo Scientific

OR321541-1G OR321541-250MG
APOSOR321541-1GEA 1060 EUR
APOSOR321541-1G APOSOR321541-250MG
(S)-4-(Piperidin-3-yl)aniline
(S)-4-(Piperidin-3-yl)aniline
Formel: C₁₁H₁₆N₂
Molecular Weight: 176.258 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20449091
CAS-Nummer: 1196713-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR