4-(Piperidin-3-yl)benzoic acid hydrochloride

Lieferant: Apollo Scientific

OR321542-1G OR321542-250MG
APOSOR321542-1GEA 498 EUR
APOSOR321542-1G APOSOR321542-250MG
4-(Piperidin-3-yl)benzoic acid hydrochloride
4-(Piperidin-3-yl)benzoic acid hydrochloride
Formel: C₁₂H₁₅NO₂·ClH
Molecular Weight: 241,714 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08460730
CAS-Nummer: 2044705-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR