(E)-N-{1-[4-(1H-Imidazol-1-yl)phenyl]ethylidene}hydroxylamine 95%

Lieferant: Apollo Scientific

OR32161-1G OR32161-5G
APOSOR32161-1GEA 339 EUR
APOSOR32161-1G APOSOR32161-5G
(E)-N-{1-[4-(1H-Imidazol-1-yl)phenyl]ethylidene}hydroxylamine 95%
(E)-N-{1-[4-(1H-Imidazol-1-yl)phenyl]ethylidene}hydroxylamine
Formel: C₁₁H₁₁N₃O
Molecular Weight: 201,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127064
CAS-Nummer: 146139-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR