Methyl isodehydroacetate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylate
OR322202-5G
OR322202-1G
APOSOR322202-5GEA
406
EUR
APOSOR322202-5G
APOSOR322202-1G
Methyl isodehydroacetate
Methyl isodehydroacetate
Formel:
C₉H₁₀O₄ Molecular Weight: 182,173 g/mol Siedepunkt: 167 °C (14 mm Hg) Schmelzpunkt: 66…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006642 CAS-Nummer: 41264-06-6 EINECS: 255-283-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...