α-chlorobenzaldoxime
Lieferant: Apollo Scientific
|
Achtung
|
OR322225-1G
OR322225-500MG
APOSOR322225-1GEA
60.9
EUR
APOSOR322225-1G
APOSOR322225-500MG
α-chlorobenzaldoxime
α-chlorobenzaldoxime
Formel:
C₇H₆ClNO Schmelzpunkt: 48…52 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02183500 CAS-Nummer: 698-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...