2-(2-Benzothiazolyl)-5-methoxyphenol

Lieferant: Apollo Scientific
Achtung

OR322356-250MG OR322356-1G
APOSOR322356-250MGEA 161 EUR
APOSOR322356-250MG APOSOR322356-1G
2-(2-Benzothiazolyl)-5-methoxyphenol
2-(2-Benzothiazolyl)-5-methoxyphenol
Formel: C₁₄H₁₁NO₂S
Molecular Weight: 257,308 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043777
CAS-Nummer: 90481-46-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR