5,6-Diaminoindazole

Lieferant: Apollo Scientific
Achtung

OR322608-100MG OR322608-250MG
APOSOR322608-100MGEA 140 EUR
APOSOR322608-100MG APOSOR322608-250MG
5,6-Diaminoindazole
5,6-Diaminoindazole
Formel: C₇H₈N₄
Molecular Weight: 148,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00022787
CAS-Nummer: 7404-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR