tert-Butyl 3-(iodomethyl)piperidine-1-carboxylate

Lieferant: Apollo Scientific

OR322791-1G OR322791-5G
APOSOR322791-1GEA 78.3 EUR
APOSOR322791-1G APOSOR322791-5G
tert-Butyl 3-(iodomethyl)piperidine-1-carboxylate
tert-Butyl 3-(iodomethyl)piperidine-1-carboxylate
Formel: C₁₁H₂₀INO₂
Molecular Weight: 325,186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09953364
CAS-Nummer: 253177-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR